Learn More
Hach Company 5500sc Standard 2 Low-Range Phosphate standard solution, 2 L - for 5500sc low-range phosphate analyzer
Catalog No. NC3240308 Shop All Hach Company Products
Encompass
Standard 2 low-range phosphate calibration solution in a 2 L bottle for use with the Hach 5500sc low-range phosphate analyzer. Use this certified standard to perform calibration and routine accuracy checks to help ensure reliable phosphate measurements.
  • Provides a low-range phosphate calibration standard for analyzer verification.
  • Packaged in a 2 L bottle for extended use and fewer changeovers.
  • Supports routine accuracy checks and instrument performance verification.
  • Ready-to-use formulation for convenient calibration procedures.
Catalog No. NC3240308 Supplier Hach Company Supplier No. 6775602
Standard 2 low-range phosphate calibration solution in a 2 L bottle for use with the Hach 5500sc low-range phosphate analyzer. Use this certified standard to perform calibration and routine accuracy checks to help ensure reliable phosphate measurements.
  • Provides a low-range phosphate calibration standard for analyzer verification.
  • Packaged in a 2 L bottle for extended use and fewer changeovers.
  • Supports routine accuracy checks and instrument performance verification.
  • Ready-to-use formulation for convenient calibration procedures.