Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, 555mm OAL
SDP
Catalog No. CG2075B03
Encompass_Preferred
Stirring Shaft, 10mm, Polished, 555mm OAL. A 10mm precision ground and polished stirrer shaft for use with PTFE bearings and CG-2091 and CG-2092 PTFE agitators. Flat ground section extends half way up shaft and permits positive seating of the PTFE locking screws.
Catalog No. CG2075B03 Supplier Chemglass Life Sciences Supplier No. CG2075B03
Stirring Shaft, 10mm, Polished, 555mm OAL. A 10mm precision ground and polished stirrer shaft for use with PTFE bearings and CG-2091 and CG-2092 PTFE agitators. Flat ground section extends half way up shaft and permits positive seating of the PTFE locking screws.