Learn More
eMolecules​ 556-18-3 | 4-Aminobenzaldehyde | ChemScene | MFCD00038137 | 121.139 | C7H7NO | 95.000 | Nc1ccc(C=O)cc1 | 25g | 572298520
Catalog No. 5000489706 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Aminobenzaldehyde | ChemScene | 556-18-3 | MFCD00038137 | 121.139 | C7H7NO | 95.000 | Nc1ccc(C=O)cc1 | 25g | 572298520
Catalog No. 50-004-89706 Supplier eMolecules​ Supplier No. 572298520
4-Aminobenzaldehyde | ChemScene | 556-18-3 | MFCD00038137 | 121.139 | C7H7NO | 95.000 | Nc1ccc(C=O)cc1 | 25g | 572298520