Learn More
Meridian Life Science 55C MMLV-RT, 10ML
SDP
Catalog No. 5000856481
Encompass_Preferred
For cDNA synthesis, a higher reaction temperature is sometimes desirable as it reduces RNA secondary structures which can inhibit reverse transcription and it minimizes nonspecific primer binding. 55C MMLV-RT is a highly thermostable reverse transcriptase with reduced RNase H activity, that allows very efficient first-strand cDNA synthesis at temperatures up to 60C. This improves the cDNA yield from difficult RNA targets, such as viral RNA genomes, that require higher temperature to denature strong RNA secondary structure.
Catalog No. 50-008-56481 Supplier Meridian Life Science Supplier No. MDX11710
For cDNA synthesis, a higher reaction temperature is sometimes desirable as it reduces RNA secondary structures which can inhibit reverse transcription and it minimizes nonspecific primer binding. 55C MMLV-RT is a highly thermostable reverse transcriptase with reduced RNase H activity, that allows very efficient first-strand cDNA synthesis at temperatures up to 60C. This improves the cDNA yield from difficult RNA targets, such as viral RNA genomes, that require higher temperature to denature strong RNA secondary structure.