Learn More
Uline Nestable Plastic Drum with Lid - 55 Gallon Open Top | Blue | 1 EA
Catalog No. NC2191400
Encompass
Protect fertilizer, ice melt and farm feed from moisture during the off-season. Drums conveniently nest when not in use. Corrosion, UV and chemical-resistant polyethylene. Lid with steel lever lock included. Stores up to 550 lbs. Two openings to pour and vent. FDA compliant. Stacking Strength 1200 lbs. Caps are not vented. Urethane rubber gasket to keep out moisture. Airtight. 1 3/4 Fine or NPS threaded opening. 1/2 Fine or NPS threaded opening.
This item is not returnable. View return policy
Catalog No. NC2191400 Supplier Uline Supplier No. S22341
Protect fertilizer, ice melt and farm feed from moisture during the off-season. Drums conveniently nest when not in use. Corrosion, UV and chemical-resistant polyethylene. Lid with steel lever lock included. Stores up to 550 lbs. Two openings to pour and vent. FDA compliant. Stacking Strength 1200 lbs. Caps are not vented. Urethane rubber gasket to keep out moisture. Airtight. 1 3/4 Fine or NPS threaded opening. 1/2 Fine or NPS threaded opening.