Learn More
Uline Plastic Drum - 55 Gallon Closed Top, Black
Catalog No. NC2393473
Encompass
Excellent for indoor or outdoor use, Corrosion free, dent-resistant polyethylene, 55 gallon stores up to 870 lbs of liquids and solids, Two 2 openings to pour and vent, FDA compliant resins
Catalog No. NC2393473 Supplier Uline Supplier No. S10757BL
Excellent for indoor or outdoor use, Corrosion free, dent-resistant polyethylene, 55 gallon stores up to 870 lbs of liquids and solids, Two 2 openings to pour and vent, FDA compliant resins