Learn More
CDF CORP 55GAL LINERS/NON STATIC/20 CS
Catalog No. NC1329957
Encompass_Preferred
DRUMSAVER™ INSERT FOR STEEL DRUMS The 5515/34.25-G4/AS-11, the anti-stat version of the popular 5515/34.25-G4 insert, is also a straight-sided, seamless, vacuum-formed insert for 55-gallon open-head steel drums. It is molded from FDA-approved low-density polyethylene. A contoured lip snaps over the top bead to prevent leakage between the insert and the drum wall, while also providing easy drum closure using a standard ring and bolt or lock assembly. INSERT DIMENSIONS Diameter: 22.5" (+0.25"/-0") Height: 34.25" (+/-0.125") Average thickness: 15 mil (+/-10%) WEIGHT 24 oz. (+/-10%) SIDE-WALL DESIGN Straight-sided TEMPERATURE PARAMETERS High (momentary): 175˚ (195˚)F Low: -40˚F ANTI-STAT Standard: MIL-B-81705-C Resistivity rating: 1011 Ω/Sq. typical CARTON 33 lb., 35"x21"x15" 20 liners per case, 10 case per pallet
Catalog No. NC1329957 Supplier CDF CORP Supplier No. 5515/34.25G4/AS11
DRUMSAVER™ INSERT FOR STEEL DRUMS The 5515/34.25-G4/AS-11, the anti-stat version of the popular 5515/34.25-G4 insert, is also a straight-sided, seamless, vacuum-formed insert for 55-gallon open-head steel drums. It is molded from FDA-approved low-density polyethylene. A contoured lip snaps over the top bead to prevent leakage between the insert and the drum wall, while also providing easy drum closure using a standard ring and bolt or lock assembly. INSERT DIMENSIONS Diameter: 22.5" (+0.25"/-0") Height: 34.25" (+/-0.125") Average thickness: 15 mil (+/-10%) WEIGHT 24 oz. (+/-10%) SIDE-WALL DESIGN Straight-sided TEMPERATURE PARAMETERS High (momentary): 175˚ (195˚)F Low: -40˚F ANTI-STAT Standard: MIL-B-81705-C Resistivity rating: 1011 Ω/Sq. typical CARTON 33 lb., 35"x21"x15" 20 liners per case, 10 case per pallet