Learn More
GREIF BROTHERS CORPORATION 55GAL TGHT HD DRM 2X2 FTNGS BL
Catalog No. NC9437818
Encompass_Preferred
Combination of an extruded body and an injection-molded bottom and cover. Produced through a unique manufacturing process, these rugged, durable drums feature virtually no variation in diameter, wall thickness and weight. These drums have been the first choice of major companies in the detergent, lubricant, pharmaceutical and agrochemical industries.
Catalog No. NC9437818 Supplier GREIF BROTHERS CORPORATION Supplier No. GP55/PTH0092 BLUE
Combination of an extruded body and an injection-molded bottom and cover. Produced through a unique manufacturing process, these rugged, durable drums feature virtually no variation in diameter, wall thickness and weight. These drums have been the first choice of major companies in the detergent, lubricant, pharmaceutical and agrochemical industries.