Learn More
Bellco Glass, Inc. 55mL Reagent Reservoir
SDP
Catalog No. 502329343
Encompass_Preferred
Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Pyramid and Trough Bottom designs promote funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution and is chemically inert for combinatorial chemistry applications. Available in Polystyrene as well.Specifications 229561000 amp 229562000 Multiple wells allow for dispensing of various reagents Autoclavable White color enhances solution visibility 55mL Reagent Reservoirs have pour spouts on corners and textured gripping areas on the sides Angled design promotes funneling of liquid to maximize retrieval Case Quantity 80
Catalog No. 50-232-9343 Supplier Bellco Glass, Inc. Supplier No. 229296000
Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Pyramid and Trough Bottom designs promote funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution and is chemically inert for combinatorial chemistry applications. Available in Polystyrene as well.Specifications 229561000 amp 229562000 Multiple wells allow for dispensing of various reagents Autoclavable White color enhances solution visibility 55mL Reagent Reservoirs have pour spouts on corners and textured gripping areas on the sides Angled design promotes funneling of liquid to maximize retrieval Case Quantity 80