Learn More
eMolecules​ 56-37-1 | Benzyltriethylammonium chloride | Combi-Blocks | MFCD00011824 | 227.780 | C13H22ClN | 97.000 | [Cl-].CC[N+](CC)(CC)Cc1ccccc1 | 5g | 290686700
Catalog No. 5000159242 Shop All eMolecules Inc. Products
Encompass_Preferred
Benzyltriethylammonium chloride | Combi-Blocks | 56-37-1 | MFCD00011824 | 227.780 | C13H22ClN | 97.000 | [Cl-].CC[N+](CC)(CC)Cc1ccccc1 | 5g | 290686700
Catalog No. 50-001-59242 Supplier eMolecules​ Supplier No. 290686700
Benzyltriethylammonium chloride | Combi-Blocks | 56-37-1 | MFCD00011824 | 227.780 | C13H22ClN | 97.000 | [Cl-].CC[N+](CC)(CC)Cc1ccccc1 | 5g | 290686700