Learn More
AdipoGen 56-Amino-TAMRA
SDP
Supplier:  AdipoGen CDXA0250M010
Encompass_Preferred
Chemical. CAS 123379-86-2. Formula C24H23N3O3. MW 401.46. Synthetic. Amino-TAMRA isomer mixture. Building block for synthesis of reactive TAMRA such isothiocyanate, iodoacetamide and maleimide. TAMRA is one of the most popular fluorophores used in various bioconjugations.
Catalog No. 50-256-1781