Learn More
AdipoGen 56-Aminofluorescein
SDP
Catalog No. 502561852
Encompass_Preferred
Chemical. CAS 27599-63-9. Formula C20H13NO5. MW 347.32. Synthetic. Two isomers are produced by the usual synthesis. Most publications imply they are interchangeable when used for the preparation of the isothiocyanate for fluorescent labeling.
Catalog No. 50-256-1852 Supplier AdipoGen Supplier No. CDXA0039G002
Chemical. CAS 27599-63-9. Formula C20H13NO5. MW 347.32. Synthetic. Two isomers are produced by the usual synthesis. Most publications imply they are interchangeable when used for the preparation of the isothiocyanate for fluorescent labeling.