Learn More
AdipoGen 56-Aminofluorescein
SDP
Catalog No. 502561851
Encompass_Preferred
Chemical. CAS 27599-63-9. Formula C20H13NO5. MW 347.32. Synthetic. Two isomers are produced by the usual synthesis. Most publications imply they are interchangeable when used for the preparation of the isothiocyanate for fluorescent labeling.
Catalog No. 50-256-1851 Supplier AdipoGen Supplier No. CDXA0039G010
Chemical. CAS 27599-63-9. Formula C20H13NO5. MW 347.32. Synthetic. Two isomers are produced by the usual synthesis. Most publications imply they are interchangeable when used for the preparation of the isothiocyanate for fluorescent labeling.