Learn More
Medchemexpress LLC HY-15940 500mg Medchemexpress, 5(6)-Carboxyfluorescein CAS:72088-94-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15940500MG
Encompass_Preferred
Medchemexpress, HY-15940 500mg 5(6)-Carboxyfluorescein CAS:72088-94-9 5(6)-Carboxyfluorescein (5(6)-FAM) is an amine-reactive pH-sensitive green fluorescent probe. 5(6)-Carboxyfluorescein (5(6)-FAM) can be used to label proteins, peptides and nucleotides. 5(6)-Carboxyfluorescein can be used for the detection of tumour areas in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1843