Learn More
Biotium 5(6)-CFDA (5-(And-6)-Carboxyfluorescein
Catalog No. 501964705
Encompass_Preferred
5(6)-CFDA (full name: 5-(and-6)-Carboxyfluorescein diacetate) is membrane-permeant and thus can be loaded into cells via incubation. Once inside cells, CFDA is hydrolyzed by intracellular esterases to carboxyfluorescein. Properties: Off-white solid soluble in DMSO; Store desiccated at 4°C; C25H16O9; MW: 460; [124387-19-5].
Catalog No. 50-196-4705 Supplier Biotium Supplier No. 51014
5(6)-CFDA (full name: 5-(and-6)-Carboxyfluorescein diacetate) is membrane-permeant and thus can be loaded into cells via incubation. Once inside cells, CFDA is hydrolyzed by intracellular esterases to carboxyfluorescein. Properties: Off-white solid soluble in DMSO; Store desiccated at 4°C; C25H16O9; MW: 460; [124387-19-5].