Learn More
AdipoGen 56-CFDA
SDP
Supplier:  AdipoGen CDXC0046M100
Encompass_Preferred
Chemical. CAS 124387-19-5. Formula C25H16O9. MW 460.39. Synthetic. CFDA is a popular, sensitive, fluorescent substrate for measuring esterase activity in live cells. It is often used in cell viability assays and "Live-Dead" assay systems, especially automated cell viability assays. The cleavage product produced upon enzymatic or chemical hydrolysis of the acetate groups is carboxyfluorescein which is well-retained inside live cells, while the substrate CFDA is membrane permeant.
Catalog No. 50-256-1514