Learn More
Abcam 5(6)-CFDA N-succinimidyl ester (CFSE), Fluorogenic esterase substrate, 500MG
Catalog No. 502705325
Encompass_Preferred
MW 557.46 g/mol, Purity >90%. Cell-permeable, fluorogenic, non-selective esterase substrate. Hydrolyzes to hydrophilic fluorescent 5(6)-CFDA (ab145278) and 5(6)-FAM (ab145342) derivatives intracellularly. Partitions with high fidelity between daughter cells for up to eight generational divisions. Tracks cell proliferation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5325 Supplier Abcam Supplier No. AB145291500MG
MW 557.46 g/mol, Purity >90%. Cell-permeable, fluorogenic, non-selective esterase substrate. Hydrolyzes to hydrophilic fluorescent 5(6)-CFDA (ab145278) and 5(6)-FAM (ab145342) derivatives intracellularly. Partitions with high fidelity between daughter cells for up to eight generational divisions. Tracks cell proliferation.
The product is subject to the following: Abcam Restricted Use Statement