Learn More
Biotium 5(6)-CARBOXYRhodamine 110-C5-Maleimide (
Catalog No. 501964830
Encompass_Preferred
CR110-C5-maleimide (full name: 5(6)-Carboxyrhodamine 110-C5-maleimide) is a maleimide derivative of the green fluorescent dye carboxyrhodamine 110. The dye has spectral properties (Ex/Em (MeOH) = 502/524 nm) similar to those of fluorescein (fluorescein-5-maleimide, for example), but the photostability of the dye is among the best of all green fluorescent dyes. Thus, the dye is a superior green fluorescent dye for labeling thiols. Properties: Orange red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C31H30N4O6; MW: 554.59.
Catalog No. 50-196-4830 Supplier Biotium Supplier No. 91029
CR110-C5-maleimide (full name: 5(6)-Carboxyrhodamine 110-C5-maleimide) is a maleimide derivative of the green fluorescent dye carboxyrhodamine 110. The dye has spectral properties (Ex/Em (MeOH) = 502/524 nm) similar to those of fluorescein (fluorescein-5-maleimide, for example), but the photostability of the dye is among the best of all green fluorescent dyes. Thus, the dye is a superior green fluorescent dye for labeling thiols. Properties: Orange red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C31H30N4O6; MW: 554.59.