Learn More
Medchemexpress LLC HY-W018025 100mg Medchemexpress, 5,6-Dihydroxyindole CAS:3131-52-0 Purity:>98%
SDP
Catalog No. 502083416
Encompass_Preferred
Medchemexpress, HY-W018025 100mg 5,6-Dihydroxyindole CAS:3131-52-0 5,6-Dihydroxyindole, a melanin precursor, has a broad-spectrum antibacterial, antifungal, antiviral, antiparasitic activity. 5,6-Dihydroxyindole has cytotoxic effects and is strongly toxic against various pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3416 Supplier Medchemexpress LLC Supplier No. HYW018025100MG
Medchemexpress, HY-W018025 100mg 5,6-Dihydroxyindole CAS:3131-52-0 5,6-Dihydroxyindole, a melanin precursor, has a broad-spectrum antibacterial, antifungal, antiviral, antiparasitic activity. 5,6-Dihydroxyindole has cytotoxic effects and is strongly toxic against various pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.