Learn More
AdipoGen 56-FAM
SDP
Supplier:  AdipoGen CDXC0045G050
Encompass_Preferred
Chemical. CAS 72088-94-9. Formula C21H12O7. MW 376.32. Synthetic. Carboxyfluorescein is commonly employed as a polar tracer and used for intracellular pH measurements. It has also been used for studying viability of various types of bacteria.
Catalog No. 50-256-1517