Learn More
AdipoGen 56-FAM N-succinimidyl ester
SDP
Catalog No. 502561556
Encompass_Preferred
Chemical. CAS 117548-22-8. Formula C25H15NO9. MW 473.4. Synthetic. Fluorescent probe and cell staining dye. Reacts with primary and secondary amine groups on proteins and other biomolecules. Cell permeable and covalently couples, via its succinimidyl group, to intracellular molecules. Due to this covalent coupling reaction fluorescent CFSE can be retained within cells for extremely long periods. Used for immunological studies to monitor the proliferation of many cell types including lymphocytes, fibroblasts and even bacteria. Has been used to prepare fluorescent derivatives of tubulin and for the preparation of a phenytoin tracer for the determination of this drug in serum and plasma by fluorescence polarization immunoassay FPIA. Favored over FITC in bioconjugations, since FAM reagents are more resistant to hydrolysis. In addition, FAM reagents require less stringent conjugation conditions and give better conjugation yields, and the resulted conjugates have superior stability.
Catalog No. 50-256-1556 Supplier AdipoGen Supplier No. CDXC0011M100
Chemical. CAS 117548-22-8. Formula C25H15NO9. MW 473.4. Synthetic. Fluorescent probe and cell staining dye. Reacts with primary and secondary amine groups on proteins and other biomolecules. Cell permeable and covalently couples, via its succinimidyl group, to intracellular molecules. Due to this covalent coupling reaction fluorescent CFSE can be retained within cells for extremely long periods. Used for immunological studies to monitor the proliferation of many cell types including lymphocytes, fibroblasts and even bacteria. Has been used to prepare fluorescent derivatives of tubulin and for the preparation of a phenytoin tracer for the determination of this drug in serum and plasma by fluorescence polarization immunoassay FPIA. Favored over FITC in bioconjugations, since FAM reagents are more resistant to hydrolysis. In addition, FAM reagents require less stringent conjugation conditions and give better conjugation yields, and the resulted conjugates have superior stability.