Learn More
AdipoGen 56-ROX
SDP
Catalog No. 502561554
Encompass_Preferred
Chemical. CAS 198978-94-8. Formula C33H30N2O5. MW 534.6. Synthetic. May be used as a fluorescent dye in nucleic acid analytical research applications.
Catalog No. 50-256-1554 Supplier AdipoGen Supplier No. CDXC0012M025
Chemical. CAS 198978-94-8. Formula C33H30N2O5. MW 534.6. Synthetic. May be used as a fluorescent dye in nucleic acid analytical research applications.