Learn More
AdipoGen 56-ROX
SDP
Supplier:  AdipoGen CDXC0012G001
Encompass_Preferred
Chemical. CAS 198978-94-8. Formula C33H30N2O5. MW 534.6. Synthetic. May be used as a fluorescent dye in nucleic acid analytical research applications.
Catalog No. 50-256-1555