Learn More
Biotium 5(6)-ROX cadaverine, TFA salt
Supplier:  Biotium 90108
Encompass_Preferred
ROX cadaverine can be coupled to activated carboxy groups or sulfonyl chlorides. Properties: Ex/Em (MeOH) = 568/595 nm Dark purple solid soluble in DMSO or DMF; Store at 4°C and protect from light; C 42 H 44 F 6 N 4 O 8; MW: 846.81
Catalog No. 50-196-4825