Learn More
AdipoGen 56-ROX N-succinimidyl ester
SDP
Catalog No. 502561552
Encompass_Preferred
Chemical. CAS 114616-32-9. Formula C37H33N3O7. MW 631.67. Synthetic. 5-and-6-Carboxy-X-rhodamine, succinimidyl ester is an amine-reactive long wavelength rhodamine dye. Derivatives made with the amine-reactive carboxy-X-rhodamine ROX, succinimidyl ester are widely used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lissamine rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red conjugates.
Catalog No. 50-256-1552 Supplier AdipoGen Supplier No. CDXC0013M025
Chemical. CAS 114616-32-9. Formula C37H33N3O7. MW 631.67. Synthetic. 5-and-6-Carboxy-X-rhodamine, succinimidyl ester is an amine-reactive long wavelength rhodamine dye. Derivatives made with the amine-reactive carboxy-X-rhodamine ROX, succinimidyl ester are widely used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lissamine rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red conjugates.