Learn More
AdipoGen 56-TAMRA
SDP
Supplier:  AdipoGen CDXC0038G002
Encompass_Preferred
Chemical. CAS 98181-63-6. Formula C25H22N2O5. MW 430.45. Synthetic. Mixed isomers of carboxytetramethylrhodamine free acid. Tetramethylrhodamine TMR is one of the fluorophores most often used for preparing peptide, protein, nucleotide and nucleic acid conjugates. TMR dyes have been widely used as acceptors for FAM fluorophores in a variety of FRET studies. The dye can be coupled to primary or secondary amines via standard peptide chemistry.
Catalog No. 50-256-1531