Learn More
AdipoGen 56-TAMRA N-SE
SDP
Catalog No. 502561538
Encompass_Preferred
Chemical. CAS 150408-83-6, 246256-50-8. Formula C29H25N3O7. MW 527.52. Synthetic. Amine-reactive form of 56-carboxytetramethylrhodamine. Conjugates yield bright, pH-insensitive orange-red fluorescence Ex/Em ~543/576nm i MeOH with good photostability. Fluorophores most often used for preparing peptide, protein, nucleotide and nucleic acid conjugates or other primary amine-containing molecules. Especially also used for fluorescent antibodies and avidin derivatives used in immunochemistry.
Catalog No. 50-256-1538 Supplier AdipoGen Supplier No. CDXC0027M025
Chemical. CAS 150408-83-6, 246256-50-8. Formula C29H25N3O7. MW 527.52. Synthetic. Amine-reactive form of 56-carboxytetramethylrhodamine. Conjugates yield bright, pH-insensitive orange-red fluorescence Ex/Em ~543/576nm i MeOH with good photostability. Fluorophores most often used for preparing peptide, protein, nucleotide and nucleic acid conjugates or other primary amine-containing molecules. Especially also used for fluorescent antibodies and avidin derivatives used in immunochemistry.