Learn More
Biotium 5(6)-Tetramethylrhodamine Maleimide
Catalog No. 501964833
Encompass_Preferred
5(6)-Carboxytetramethylrhodamine maleimide has C6-linker that enhances the fluorescence after the dye is conjugated to proteins. The product is a direct replacement for 5(6)-carboxytetramethylrhodamine C5-maleimide (91040).Properties: Ex/Em (MeOH) = 540/565 nm; Red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C35H36N4O6; MW: 608.66.
Catalog No. 50-196-4833 Supplier Biotium Supplier No. 91054
5(6)-Carboxytetramethylrhodamine maleimide has C6-linker that enhances the fluorescence after the dye is conjugated to proteins. The product is a direct replacement for 5(6)-carboxytetramethylrhodamine C5-maleimide (91040).Properties: Ex/Em (MeOH) = 540/565 nm; Red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C35H36N4O6; MW: 608.66.