Learn More
Medchemexpress LLC HY-15398A 5mg Medchemexpress, 5,6-trans-Vitamin D3 CAS:22350-41-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15398A5MG
Encompass_Preferred
Medchemexpress, HY-15398A 5mg 5,6-trans-Vitamin D3 CAS:22350-41-0 5,6-trans-Vitamin D3 (5,6-trans-Cholecalciferol;5,6-trans-Colecalciferol) is a photoproduct of vitamin D3. Vitamin D3 is a naturally occuring form of vitamin D. Vitamin D3 induces cell differentiation and prevents proliferation of cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1716