Learn More
Wells Lamont Industrial™ 5600 Cut Resistant Glove
Catalog No. 18063990 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-063-990 X-Small
18-063-991 Small
18-063-992 Medium
18-063-993 Large
18-063-994 X-Large
18-063-995 2X-Large
18-063-996 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-063-990 Supplier Wells Lamont Industrial™ Supplier No. 5600XS
Only null left
Add to Cart
Add to Cart
5600 ANSI A8 Level cut resistant glove. Each size is a different color for easy identification.
The 15g gauge 5600 series glove is unique. Not only can the color be spotted, but it provides dexterity and cut protection for the employee on the line.
  • Knitted with high-performance fibers and wire to achieve the level of cut resistance needed for protection without sacrificing dexterity.
  • New and improved cut resistance is now an ANSI A8 level.
  • Not only will the thin glove protect against possible lacerations when working with knives it allows movement to reduce carpal tunnel or connect tissue disorders.
  • Safety awareness is easily visible with the bright color selection.
  • The color by size is easy for identification plus the easy visibility assures employees are wearing hand protection.
  • Select the 5600 color series gloves for more dexterity and increased awareness with the bright color selection it offers.
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A8
Hand Ambidextrous
Latex Free Yes
Material High Performance Fibers
Product Type Cut-resistant Gloves
Size X-Small
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Straight