Learn More
eMolecules​ 5674-01-1 | 2-Methylprop-2-en-1-ylmagnesium chloride, 0.50 M in THF | Rieke Metals | MFCD00674003 | 114.860 | C4H7ClMg | 0.000 | [Cl-].CC(=C)C[Mg+] | 50ml | 415758820
Catalog No. NC3858460 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Methylprop-2-en-1-ylmagnesium chloride, 0.50 M in THF | Rieke Metals | 5674-01-1 | MFCD00674003 | 114.860 | C4H7ClMg | 0.000 | [Cl-].CC(=C)C[Mg+] | 50ml | 415758820
Catalog No. NC3858460 Supplier eMolecules​ Supplier No. 415758820
2-Methylprop-2-en-1-ylmagnesium chloride, 0.50 M in THF | Rieke Metals | 5674-01-1 | MFCD00674003 | 114.860 | C4H7ClMg | 0.000 | [Cl-].CC(=C)C[Mg+] | 50ml | 415758820