Learn More
Aat Bioquest 5(6)-FAM ethylenediamine
SDP
Catalog No. 502979218
Encompass_Preferred
5(6)-FAM ethylenediamine is an excellent building block for developing fluoresceinated bioconjugates.
Catalog No. 50-297-9218 Supplier Aat Bioquest Supplier No. 123
5(6)-FAM ethylenediamine is an excellent building block for developing fluoresceinated bioconjugates.