Learn More
Aat Bioquest 5(6)-FAM, SE [5-(and-6)-Carboxyfluorescein, succinimidyl ester] [CAS 117548-22-8]
SDP
Catalog No. 502979127
Encompass_Preferred
Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.
Catalog No. 50-297-9127 Supplier Aat Bioquest Supplier No. 112
Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.