Learn More
Medchemexpress LLC HY-N1970 5mg Medchemexpress, 5,7-Dihydroxychromone CAS:31721-94-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19705MG
Encompass_Preferred
Medchemexpress, HY-N1970 5mg 5,7-Dihydroxychromone CAS:31721-94-5 5,7-Dihydroxychromone, the extract of Cudrania tricuspidata, activates Nrf2/ARE signal and exerts neuroprotective effects against 6-hydroxydopamine (6-OHDA)-induced oxidative stress and apoptosis. 5,7-Dihydroxychromone inhibits the expression of activated caspase-3 and caspase-9 and cleaved PARP in 6-OHDA-induced SH-SY5Y cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2855