Learn More
Medchemexpress LLC HY-N5011 25mg Medchemexpress, 5,7-Dimethoxyflavone CAS:21392-57-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN501125MG
Encompass_Preferred
Medchemexpress, HY-N5011 25mg 5,7-Dimethoxyflavone CAS:21392-57-4 5,7-Dimethoxyflavone is one of the major components of Kaempferia parviflora, has anti-obesity, anti-inflammatory, and antineoplastic effects. 5,7-Dimethoxyflavone inhibits cytochrome P450 (CYP) 3As. 5,7-Dimethoxyflavone is also a potent Breast Cancer Resistance Protein (BCRP) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2936