Learn More
Sino Biological Recombinant Mouse CD47 Protein (rFc Tag)
Supplier:  Sino Biological 57231M31H200
Encompass_Preferred
A DNA sequence encoding the mouse Cd47 (XP_006521874.1) (Met1-Lys140) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-3187