Learn More
Sino Biological Recombinant Mouse Neuropilin-2/NRP2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 57465M08H100
Encompass_Preferred
A DNA sequence encoding the mouse NRP2 (NP_001070871.1) (Met1-Pro864) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3191