Learn More
Sino Biological Recombinant Mouse GITR/TNFRSF18/AITR Protein (mFc Tag)
Supplier:  Sino Biological 57511M05H100
Encompass_Preferred
A DNA sequence encoding the mouse TNFRSF18 (NP_033426.1) (Met1-His153) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-162-3193