Learn More
Ace Glass, Inc. 575mm x 14mm OD porosity A straight sparger tube, 1/2" connection top
SDP
Catalog No. 50978746
Encompass_Preferred
575mm X 14mm O.D. straight sparger tube with a 145-174 micron fritted disc at the bottom, and a 1/2 I.D. hose connection tubing at the top. Will fit inside #15 Ace-Threds.
Catalog No. 50-978-746 Supplier Ace Glass, Inc. Supplier No. 645321
575mm X 14mm O.D. straight sparger tube with a 145-174 micron fritted disc at the bottom, and a 1/2 I.D. hose connection tubing at the top. Will fit inside #15 Ace-Threds.