Learn More
Ace Glass, Inc. 575mm x 14mm OD porosity B straight sparger tube, #7Ace-Thred top, 5029-10 nylon bushing and FETFE O-ring included
SDP
Catalog No. 50978748
Encompass_Preferred
575mm X 14mm O.D. straight sparger tube with a 70-100micron fritted disc at the bottom, and a #7 Ace-Thred for 7mm O.D. tubing at the top. Supplied with nylon bushing and FETFE o-ring. Will fit inside #15 Ace-Threds.
Catalog No. 50-978-748 Supplier Ace Glass, Inc. Supplier No. 6453124
575mm X 14mm O.D. straight sparger tube with a 70-100micron fritted disc at the bottom, and a #7 Ace-Thred for 7mm O.D. tubing at the top. Supplied with nylon bushing and FETFE o-ring. Will fit inside #15 Ace-Threds.