Learn More
Ace Glass, Inc. Acrylic safety shield and polyethylene safety cap for Michel-Miller chromatography columns. Cap is 57mm ID and fits#11 threaded columns. The shield is 17cm long.
SDP
Catalog No. 5097282
Encompass_Preferred
Catalog No. 50-972-82 Supplier Ace Glass, Inc. Supplier No. 5798235