Learn More
Ace Glass, Inc. Acrylic safety shield and aluminum safety cap for Michel-Miller chromatography columns. Cap is 57mm ID and fits#11 threaded columns. The shield is 33.5cm long.
SDP
Catalog No. 5097260
Encompass_Preferred
Catalog No. 50-972-60 Supplier Ace Glass, Inc. Supplier No. 579836