Learn More
Ace Glass, Inc. Acrylic safety shield and aluminum safety cap for Michel-Miller chromatography columns. Cap is 57mm ID and fits#15 threaded columns. The shield is 45cm long.
SDP
Supplier:  Ace Glass, Inc. 579852
Encompass_Preferred
57mm I.D. x 45cm L Acrylic Safety Shield with aluminum cap, for use with #15 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.
Catalog No. 50-978-428