Learn More
Ace Glass, Inc. Acrylic safety shield and polyethylene safety cap for Michel-Miller chromatography columns. Cap is 57mm ID and fits#15 threaded columns. The shield is 45cm long.
SDP
Supplier:  Ace Glass, Inc. 5798252
Encompass_Preferred
Catalog No. 50-972-85