Learn More
Ace Glass, Inc. Acrylic safety shield and polyethylene safety cap for Michel-Miller chromatography columns. Cap is 57mm ID and fits#25 threaded columns. The shield is 60cm long.
SDP
Catalog No. 50978435
Encompass_Preferred
57mm I.D. x 60cm L Acrylic Safety Shield with polyethylene safety cap, for use with #25 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.
Catalog No. 50-978-435 Supplier Ace Glass, Inc. Supplier No. 5798258
57mm I.D. x 60cm L Acrylic Safety Shield with polyethylene safety cap, for use with #25 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.