Learn More
Ace Glass, Inc. Acrylic safety shield and aluminum safety cap for Michel-Miller chromatography columns. Cap is 57mm ID and fits#50 threaded columns. The shield is 60cm long.
SDP
Catalog No. 5097270
Encompass_Preferred
Catalog No. 50-972-70 Supplier Ace Glass, Inc. Supplier No. 579858