Learn More
Chemglass Life Sciences 57mm Ground Flat Flange, Tube ID: 52mm, Tube OD: 57mm
SDP
Catalog No. CG13502
Encompass_Preferred
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 57mm Ground Flat Flange, Tube ID: 52mm, Tube OD: 57mm.
Catalog No. CG13502 Supplier Chemglass Life Sciences Supplier No. CG13502
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 57mm Ground Flat Flange, Tube ID: 52mm, Tube OD: 57mm.