Learn More
Chemglass Life Sciences Clamp, Horseshoe Style, 57mm Flange
SDP
Catalog No. CG13902
Encompass_Preferred
Clamp, Horseshoe Style, 57mm Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.
Catalog No. CG13902 Supplier Chemglass Life Sciences Supplier No. CG13902
Clamp, Horseshoe Style, 57mm Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.