Learn More
Ace Glass, Inc. 57mm ID polyethylene shield cap fitting #11 threaded chromatography columns.
SDP
Supplier:  Ace Glass, Inc. 5798133
Encompass_Preferred
57mm I.D. Polyethylene Shield Cap, for use with acrylic safety shield for #11 threaded Michel-Miller Chromatography Columns. Safety shield insures safe operation at pressures to 300 psi.
Catalog No. 50-978-431