Learn More
Perkin Elmer US LLC 5.8 kVA Power Conditioner for AAnalyst 600/800/ELAN 6x00/9000 RF Generator Side- 60 Hz
Supplier:  Perkin Elmer US LLC N9307511
Encompass_Preferred
24.2A, 208/240V Input, 120/208/ Output Isolation, Filtering and Surge Suppression L6-30P Input plug, (1)L6-30R, (4)5-20R Output Receptacles Damaged or compromised components, disrupted processes, lack of reliability they all add up to frustration, broken schedules and costly downtime. The problem is caused by a host of power disturbances some visible and many invisible that threaten your equipments operation every year. Power disturbances are simply those large and small variations in the quality of the electric power you use day in and day out. Some come from your local utility company, but most are created within your own facility as a result of the distribution and use of electrical power. These variations include high-energy voltage transients, sags and swells, electrical noise, and common mode voltage, as well as the power outages everyone witnesses when the lights go out.
Catalog No. 50-211-3130
Add to Cart
Request a Quote